5-Norbornene-2-endo,3-endo-dimethanol ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Bicyclo[2.2.1]hept-5-ene-2-endo,3-endo-dimethanol, 5-endo,6-endo-Bis(hydroxymethyl)-2-norbornene

N0768-1G N0768-5G
TCIAN0768-1GEA 870 SEK
TCIAN0768-1G TCIAN0768-5G
5-Norbornene-2-endo,3-endo-dimethanol ≥98.0%
5-Norbornene-2-endo,3-endo-dimethanol
Formel: C₉H₁₄O₂
MW: 154,21 g/mol
Kokpunkt: 130°C
MDL Number: MFCD00674044
CAS nummer: 699-97-8

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 84.0 to 88.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR