Potassium bisnonafluoro-1-butanesulphonimidate ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Bis(nonafluoro-1-butanesulphonyl)amine Potassium Salt, Nonafluoro-1-butanesulphonimide Potassium Salt, Bisnonafluoro-1-butanesulphonimide Potassium Salt, Potassium Nonafluoro-1-butanesulphonimidate

N0712-1G N0712-5G
TCIAN0712-1GEA 685 SEK
TCIAN0712-1G TCIAN0712-5G
Potassium bisnonafluoro-1-butanesulphonimidate ≥98.0%
Potassium bisnonafluoro-1-butanesulphonimidate
Formel: C₈HF₁₈NO₄S₂
MW: 619,28 g/mol
MDL Number: MFCD06200794
CAS nummer: 129135-87-1

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR