α-linolensyra 99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: ALA, (9Z,12Z,15Z)-Octadeca-9,12,15-trienoic acid, Linolenic acid, 9Z,12Z,15Z-Octadecatrienoic acid, cis,cis,cis-9,12,15-Octadecatrienoic acid
302820010 302820050 302825000
ACRO302820010EA 4175 SEK
ACRO302820010 ACRO302820050 ACRO302825000
α-linolensyra 99%
α-linolensyra
Formel: C₁₈H₃₀O₂
Kokpunkt: 230…232 °C (1 mmHg)
Smältpunkt: –11 °C
Densitet: 0,91 g/cm³
Flampunkt: >112 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00065720
CAS nummer: 463-40-1
EINECS: 207-334-8
Merck Index: 12,05530
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4790 to 1.4810 (20°C, 589 nm)
GC ≥98.5 %