3-(2-Naphthyl)-L-alanine ≥97.0%

Leverantör: TCI EUROPE

Synonyms: (S)-alpha-Amino-2-naphthalenepropionic Acid, (S)-2-Amino-3-(naphthalen-2-yl)propanoic acid, H-3-(2-Naphthyl)-Ala-OH

N0646-1G N0646-5G
TCIAN0646-1GEA 1500 SEK
TCIAN0646-1G TCIAN0646-5G
3-(2-Naphthyl)-L-alanine ≥97.0%
3-(2-Naphthyl)-L-alanine
Formel: C₁₃H₁₃NO₂
MW: 215,25 g/mol
MDL Number: MFCD00066087
CAS nummer: 58438-03-2

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Specific rotation [a]20/D -10.0 to -12.0 deg (C=1, 1mol/L HCl)
Solubility in 1mol/L NaOH almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR