2-(5-Norbornene-2,3-dicarboximido)-1,1,3,3-tetramethyluronium tetrafluoroborate coupling agent for peptides synthesis

Leverantör: TCI EUROPE
Fara

Synonyms: TNTU, 2-(1,3-Dioxo-3a,4,7,7a-tetrahydro-1H-4,7-methanoisoindol-2(3H)-yl)-1,1,3,3-tetramethyluronium tetrafluoroborate

N0634-25G N0634-5G
TCIAN0634-25GEA 1750 SEK
TCIAN0634-25G TCIAN0634-5G
2-(5-Norbornene-2,3-dicarboximido)-1,1,3,3-tetramethyluronium tetrafluoroborate coupling agent for peptides synthesis
2-(5-Norbornene-2,3-dicarboximido)-1,1,3,3-tetramethyluronium tetrafluoroborate
Formel: C₁₄H₂₀BF₄N₃O₃
MW: 365,14 g/mol
MDL Number: MFCD00167753
CAS nummer: 125700-73-4
UN: 1759
ADR: 8,III

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 96.0 %
Solubility in N,N-DMF almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR