2-Nitro-p-xylylene glycol ≥95.0%
Leverantör: TCI
Synonyms:
(2-Nitro-1,4-phenylene)dimethanol
N0519-5G
N0519-1G
TCIAN0519-5GEA
2725
SEK
TCIAN0519-5G
TCIAN0519-1G
2-Nitro-p-xylylene glycol ≥95.0%
2-Nitro-p-xylylene glycol
Formel:
C₈H₉NO₄ MW: 183.16 g/mol |
MDL Number:
MFCD00024286 CAS nummer: 23222-97-1 EINECS: 245-503-8 |
|
Specifikation testresultat
Purity (GC) | min. 95.0 % |
Melting point | 92.0 to 97.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...