3-Nitropyrrole ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 3-Nitro-1H-pyrrole

N0502-1G N0502-5G
TCIAN0502-1GEA 1625 SEK
TCIAN0502-1G TCIAN0502-5G
3-Nitropyrrole ≥98.0%
3-Nitropyrrole
Formel: C₄H₄N₂O₂
MW: 112,09 g/mol
MDL Number: MFCD00059714
CAS nummer: 5930-94-9

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 98.0 to 101.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR