Nicotinic acid N-oxide ≥98.0%

Leverantör: TCI
Varning

Synonyms: Oxiniacic Acid, 3-Carboxypyridine 1-oxide

N0497-25G N0497-500G
TCIAN0497-25GEA 325 SEK
TCIAN0497-25G TCIAN0497-500G
Nicotinic acid N-oxide ≥98.0%
Nicotinic acid N-oxide
Formel: C₆H₅NO₃
MDL Number: MFCD00006201
CAS nummer: 2398-81-4
EINECS: 219-265-0
Merck Index: 14,06941

Order Now

Specifikation testresultat

Appearance White to Light yellow to Dark green powder to crystal
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR