3,3',3''-Nitrilotripropionic acid ≥99.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3,3',3''-Nitrilotripropanoic acid

N0409-10G N0409-25G
TCIAN0409-10GEA 1925 SEK
TCIAN0409-10G TCIAN0409-25G
3,3',3''-Nitrilotripropionic acid ≥99.0%
3,3',3''-Nitrilotripropionic acid
Formel: C₉H₁₅NO₆
MW: 233,22 g/mol
MDL Number: MFCD00055827
CAS nummer: 817-11-8

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 99.0 %
Solubility in 1mol/L NaOH almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR