Sodium-2-nitroaniline-4-sulphonate ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
2-Nitroaniline-4-sulphonic acid Sodium Salt, Sodium 4-Amino-3-nitrobenzenesulphonate, 4-Amino-3-nitrobenzenesulphonic acid Sodium Salt
N0371-25G
N0371-500G
N0371-100G
TCIAN0371-25GEA
550
SEK
TCIAN0371-25G
TCIAN0371-500G
TCIAN0371-100G
Sodium-2-nitroaniline-4-sulphonate ≥98.0%
Sodium-2-nitroaniline-4-sulphonate
Formel:
C₆H₆N₂O₅S MW: 240,16 g/mol |
MDL Number:
MFCD00070599 CAS nummer: 5042-33-1 EINECS: 225-735-6 |
Specifikation testresultat
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Ion exchange titration) | min. 95.0 % (calcd.on anh.substance) |
Solubility in Water | very faint turbidity |
Drying loss | max. 2.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...