Dimethyl-2,6-naphthalenedicarboxylate ≥99.0%
Leverantör: TCI EUROPE
Synonyms:
Dimethyl 2,6-naphthalenedicarboxylate, 2,6-Naphthalenedicarboxylic Acid Dimethyl Ester
N0364-25G
N0364-5G
TCIAN0364-25GEA
1400
SEK
TCIAN0364-25G
TCIAN0364-5G
Dimethyl-2,6-naphthalenedicarboxylate ≥99.0%
Dimethyl-2,6-naphthalenedicarboxylate
Formel:
C₁₄H₁₂O₄ MW: 244,25 g/mol |
MDL Number:
MFCD00004100 CAS nummer: 840-65-3 EINECS: 212-661-4 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 190.0 to 193.0 deg-C |
Solubility in hot Toluene | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...