N-(2-Nitrophenylsulphenyl)-S-benzyl-L-cysteine dicyclohexylammonium salt ≥99.0%
Leverantör: TCI EUROPE
Synonyms:
N-(2-Nitrophenylthio)-S-benzyl-L-cysteine Dicyclohexylammonium Salt, Nps-Cys(Bzl)-OH.DCHA, N-Nps-S-benzyl-L-cysteine Dicyclohexylammonium Salt
N0361-1G
N0361-5G
TCIAN0361-1GEA
375
SEK
TCIAN0361-1G
TCIAN0361-5G
N-(2-Nitrophenylsulphenyl)-S-benzyl-L-cysteine dicyclohexylammonium salt ≥99.0%
N-(2-Nitrophenylsulphenyl)-S-benzyl-L-cysteine dicyclohexylammonium salt
Formel:
C₁₆H₁₆N₂O₄S₂ MW: 545,76 g/mol |
MDL Number:
MFCD00038490 CAS nummer: 7675-65-2 |
Specifikation testresultat
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (Neutralization titration) | min. 99.0 % |
Specific rotation [a]20/D | -42.0 to -45.0 deg (C=0.7, DMF) |
Solubility in N,N-DMF | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...