4-Nitrosoresorcinol-1-monomethyl ether ≥98.0%

Leverantör: TCI
Varning

Synonyms: Coniferron, 2-Hydroxy-4-methoxy-1-nitrosobenzene, 5-Methoxy-2-nitrosophenol, 4-Methoxy-1,2-benzoquinone 1-Monoxime

N0271-1G N0271-25G
TCIAN0271-1GEA 215 SEK
TCIAN0271-1G TCIAN0271-25G
4-Nitrosoresorcinol-1-monomethyl ether ≥98.0%
4-Nitrosoresorcinol-1-monomethyl ether
Formel: C₇H₇NO₃
MW: 153.14 g/mol
MDL Number: MFCD00059952
CAS nummer: 13895-38-0
UN: 1325
ADR: 4.1 III

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in hot EtOH almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR