5-Dimethylamino-2-nitrosophenol hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: 2-Nitroso-5-dimethylaminophenol Hydrochloride

N0261-25G N0261-5G
TCIAN0261-25GEA 5900 SEK
TCIAN0261-25G TCIAN0261-5G
5-Dimethylamino-2-nitrosophenol hydrochloride ≥98.0% (by HPLC, titration analysis)
5-Dimethylamino-2-nitrosophenol hydrochloride
Formel: C₈H₁₀N₂O₂
MW: 202,64 g/mol
MDL Number: MFCD00060162
CAS nummer: 41317-10-6

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR