Trisodium naphthalene-1,3,6-trisulphonate hydrate >65.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Naphthalene-1,3,6-trisulphonic acid Trisodium Salt
N0021-25G
N0021-500G
N0021-100G
TCIAN0021-25GEA
270
SEK
TCIAN0021-25G
TCIAN0021-500G
TCIAN0021-100G
Trisodium naphthalene-1,3,6-trisulphonate hydrate >65.0%
Trisodium naphthalene-1,3,6-trisulphonate hydrate
Formel:
C₁₀H₈O₉S₃·xH₂O MW: 434,29 g/mol |
MDL Number:
MFCD00003989 CAS nummer: 5182-30-9 EINECS: 225-962-0 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 65.0 area% |
Purity (Ion exchange titration) | min. 95.0 % (calcd.on anh.substance) |
Water | max. 12.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...