Methyl 1,4-Dioxaspiro[4.5]decane-8-carboxylate
Leverantör: TCI EUROPE
Synonyms:
Methyl 1,4-dioxaspiro[4.5]decane-8-carboxylate, 1,4-Dioxaspiro[4.5]decane-8-carboxylic acid methyl ester
M3273-1G
M3273-5G
TCIAM3273-1GEA
765
SEK
TCIAM3273-1G
TCIAM3273-5G
Methyl 1,4-Dioxaspiro[4.5]decane-8-carboxylate
Methyl 1,4-Dioxaspiro[4.5]decane-8-carboxylate
Formel:
C₁₀H₁₆O₄ MW: 200,23 g/mol Kokpunkt: 140°C |
MDL Number:
MFCD13195278 CAS nummer: 26845-47-6 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...