4-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)benzoic acid ≥98.0% (by GC, total nitrogen)
Leverantör: TCI EUROPE
Varning
Synonyms: N-(4-Carboxyphenyl)maleimide, 4-Maleimidobenzoic acid, 4-(2,5-Dioxo-2,5-dihydro-pyrrol-1-yl)-benzoicacid
M3257-1G M3257-5G
TCIAM3257-1GEA 765 SEK
TCIAM3257-1G TCIAM3257-5G
4-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)benzoic acid ≥98.0% (by GC, total nitrogen)
4-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)benzoic acid
Formel: C₁₁H₇NO₄
MW: 217,18 g/mol
Storage Temperature: Frys
MDL Number: MFCD00458571
CAS nummer: 17057-04-4
EINECS: 241-117-9
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 237.0 to 241.0 deg-C