1-[(4-Methoxyphenyl)ethynyl]-4-n-pentylbenzene ≥98.0%

Leverantör: TCI

M3230-25G M3230-5G
TCIAM3230-25GEA 4175 SEK
TCIAM3230-25G TCIAM3230-5G
1-[(4-Methoxyphenyl)ethynyl]-4-n-pentylbenzene ≥98.0%
1-[(4-Methoxyphenyl)ethynyl]-4-n-pentylbenzene
Formel: C₂₀H₂₂O
Smältpunkt: 45°C
MDL Number: MFCD00187208
CAS nummer: 39969-28-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f11b66b5fd991d3646a29e495654bfe5c202be8&vid=8fd0d83d4c57d0f8a1823ffb375f8bd83bd2bd64&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR