1-Methylcyclopentyl methacrylate ≥98.0% stabilised

Leverantör: TCI

Synonyms: Methacrylic acid 1-Methylcyclopentyl ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

M3228-1G M3228-5G
TCIAM3228-1GEA 510 SEK
TCIAM3228-1G TCIAM3228-5G
1-Methylcyclopentyl methacrylate ≥98.0% stabilised
1-Methylcyclopentyl methacrylate
Formel: C₁₀H₁₆O₂
MW: 168.24 g/mol
CAS nummer: 178889-45-7

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR