2-Methyl-4,6-bis[(n-octylthio)methyl]phenol 97,0 GC_ANALYSMETOD

Leverantör: TCI

Synonyms: 2-Methyl-4,6-bis(n-octylsulphanylmethyl)phenol, 2-Methyl-4,6-bis((octylthio)methyl)phenol

M3227-25G M3227-500G
TCIAM3227-25GEA 245 SEK
TCIAM3227-25G TCIAM3227-500G
2-Methyl-4,6-bis[(n-octylthio)methyl]phenol 97,0 GC_ANALYSMETOD
2-Methyl-4,6-bis[(n-octylthio)methyl]phenol
Formel: C₂₅H₄₄OS₂
MW: 424.75 g/mol
MDL Number: MFCD00134699
CAS nummer: 110553-27-0
EINECS: 402-860-6

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR