4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-[(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl]cyclohexanecarboxylic acid, 4-((2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl)cyclohexanecarboxylic acid, N-(4-Carboxycyclohexylmethyl)maleimide
M3218-1G
M3218-5G
TCIAM3218-1GEA
1575
SEK
TCIAM3218-1G
TCIAM3218-5G
4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid ≥98.0%
4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid
Formel:
C₁₂H₁₅NO₄ MW: 237,26 g/mol Smältpunkt: 91°C Storage Temperature: Frys |
MDL Number:
MFCD03093740 CAS nummer: 64987-82-2 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % (total of isomer) |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...