Methoxy(dimethyl)-n-octylsilane 95,0 GC_ANALYSMETOD
Leverantör: TCI
|
Varning
|
Synonyms:
Methoxydimethyl(octyl)silane
M3217-5ML
M3217-25ML
TCIAM3217-5MLEA
445
SEK
TCIAM3217-5ML
TCIAM3217-25ML
Methoxy(dimethyl)-n-octylsilane 95,0 GC_ANALYSMETOD
Methoxy(dimethyl)-n-octylsilane
Formel:
C₁₁H₂₆OSi MW: 202.41 g/mol Flampunkt: 83°C |
MDL Number:
MFCD00043322 CAS nummer: 93804-29-6 EINECS: 298-404-7 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...