1-Methylimidazole trifluoromethanesulphonate ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI
Varning

Synonyms: 1-Methylimidazolium trifluoromethanesulphonate, 1-Methylimidazole triflate, 1-Methylimidazolium triflate

M3214-5G M3214-25G
TCIAM3214-5GEA 1075 SEK
TCIAM3214-5G TCIAM3214-25G
1-Methylimidazole trifluoromethanesulphonate ≥98.0% (by HPLC, titration analysis)
1-Methylimidazole trifluoromethanesulphonate
Formel: C₅H₇F₃N₂O₃S
Smältpunkt: 88°C
MDL Number: MFCD18251289
CAS nummer: 99257-94-0

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR