1-Methylimidazole hydrobromide ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1-Methyl-1H-imidazole hydrobromide, 1-Methylimidazolium bromide
M3212-5G
M3212-25G
TCIAM3212-5GEA
230
SEK
TCIAM3212-5G
TCIAM3212-25G
1-Methylimidazole hydrobromide ≥98.0% (by HPLC, titration analysis)
1-Methylimidazole hydrobromide
Formel:
C₄H₆N₂·HBr MW: 163,02 g/mol |
MDL Number:
MFCD18251295 CAS nummer: 101023-58-9 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...