Malonic acid dihydrazide ≥93.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Malonohydrazide
M3206-5G
M3206-25G
TCIAM3206-5GEA
525
SEK
TCIAM3206-5G
TCIAM3206-25G
Malonic acid dihydrazide ≥93.0%
Malonic acid dihydrazide
Formel:
C₃H₈N₄O₂ MW: 132,12 g/mol |
MDL Number:
MFCD00041268 CAS nummer: 3815-86-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 93.0 % |
Melting point | 156.0 to 160.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...