4-(4-Methylphenylazo)phenol ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI
Varning

M3204-1G M3204-5G
TCIAM3204-1GEA 845 SEK
TCIAM3204-1G TCIAM3204-5G
4-(4-Methylphenylazo)phenol ≥98.0% (by HPLC, titration analysis)
4-(4-Methylphenylazo)phenol
Formel: C₁₃H₁₂N₂O
CAS nummer: 2497-33-8

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 151.0 to 155.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR