3-Methyl-1H-1,2,4-triazole ≥98.0%
Leverantör: TCI EUROPE
Varning
M3178-1G M3178-5G
TCIAM3178-1GEA 485 SEK
TCIAM3178-1G TCIAM3178-5G
3-Methyl-1H-1,2,4-triazole ≥98.0%
3-Methyl-1H-1,2,4-triazole
Formel: C₃H₅N₃
MW: 83,0900 g/mol
Kokpunkt: 265°C
MDL Number: MFCD00233931
CAS nummer: 7170-01-6
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 94.0 to 98.0 deg-C