5-Methoxyoxindole ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
5-Methoxy-2-indolinone, 5-Methoxyindolin-2-one
M3157-200MG
M3157-1G
TCIAM3157-200MGEA
685
SEK
TCIAM3157-200MG
TCIAM3157-1G
5-Methoxyoxindole ≥98.0%
5-Methoxyoxindole
Formel:
C9H9NO2 MW: 163,1733 g/mol |
MDL Number:
MFCD01860251 CAS nummer: 7699-18-5 |
Specifikation testresultat
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 152.0 to 156.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...