3-(Methoxycarbonyl)bicyclo[1.1.1]pentane-1-carboxylic acid ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Bicyclo[1.1.1]pentane-1,3-dicarboxylic acid 1-Monomethyl ester

M3149-200MG M3149-1G
TCIAM3149-200MGEA 950 SEK
TCIAM3149-200MG TCIAM3149-1G
3-(Methoxycarbonyl)bicyclo[1.1.1]pentane-1-carboxylic acid ≥97.0%
3-(Methoxycarbonyl)bicyclo[1.1.1]pentane-1-carboxylic acid
Formel: C₈H₁₀O₄
MW: 170,16 g/mol
MDL Number: MFCD20638314
CAS nummer: 83249-10-9

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 138.0 to 142.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR