2-Maleimidoacetic acid ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)acetic acid

M3143-50MG M3143-250MG
TCIAM3143-50MGEA 510 SEK
TCIAM3143-50MG TCIAM3143-250MG
2-Maleimidoacetic acid ≥98.0% (by HPLC, titration analysis)
2-Maleimidoacetic acid
Formel: C₆H₅NO₄
MW: 155,11 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00457254
CAS nummer: 25021-08-3

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 114.0 to 118.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR