Methacrylic anhydride ≥97.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: Methacrylic Anhydride [stabilized with 1,1,3-Tris(3-tert-butyl-4-hydroxy-6-methylphenyl)butane]
M3139-100G M3139-500G
TCIAM3139-100GEA 645 SEK
TCIAM3139-100G TCIAM3139-500G
Methacrylic anhydride ≥97.0% (by GC, titration analysis)
Methacrylic anhydride
Formel: C₈H₁₀O₃
MW: 154,165 g/mol
Kokpunkt: 205°C
Flampunkt: 83°C
Storage Temperature: Kylskåp
MDL Number: MFCD00008586
CAS nummer: 760-93-0
EINECS: 212-084-8
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Morpholine Method) min. 97.0 %