2-(3-Methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)anisol, 3-Methoxyphenylboronic acid pinacol ester

M3124-1G M3124-5G
TCIAM3124-1GEA 335 SEK
TCIAM3124-1G TCIAM3124-5G
2-(3-Methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)
2-(3-Methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formel: C₁₃H₁₉BO₃
MW: 234,10 g/mol
MDL Number: MFCD05863915
CAS nummer: 325142-84-5

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR