1-Methyl-3-(4-sulphobutyl)imidazolium bis(trifluoromethanesulphonyl)imide ≥95.0%

Leverantör: TCI EUROPE
Varning

M3119-1G M3119-5G
TCIAM3119-1GEA 415 SEK
TCIAM3119-1G TCIAM3119-5G
1-Methyl-3-(4-sulphobutyl)imidazolium bis(trifluoromethanesulphonyl)imide ≥95.0%
1-Methyl-3-(4-sulphobutyl)imidazolium bis(trifluoromethanesulphonyl)imide
Formel: C₁₀H₁₅F₆N₃O₇S₃
MW: 499,41 g/mol
CAS nummer: 909390-59-6

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR