Mdivi -1 ≥95.0%
Leverantör: TCI EUROPE
Synonyms: Mdivi-1, 3-(2,4-Dichloro-5-methoxyphenyl)-2-mercaptoquinazolin-4(3H)-one, 3-(2,4-Dichloro-5-methoxyphenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
M3108-10MG M3108-50MG
TCIAM3108-10MGEA 1550 SEK
TCIAM3108-10MG TCIAM3108-50MG
Mdivi -1 ≥95.0%
Mdivi -1
Formel: C₁₅H₁₀Cl₂N₂O₂S
MW: 353,2 g/mol
Storage Temperature: Frys
MDL Number: MFCD00974506
CAS nummer: 338967-87-6
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 7.50 to 8.50 %