Methyl-PEG24-Azide ≥95.0%
Leverantör: TCI EUROPE
Synonyms:
mPEG24-Azide, O-(2-Azidoethyl)-O'-methyltricosaethylene Glycol, mPEG24-N3, 73-Azido-2,5,8,11,14,17,20,23,26,29,32,35,38,41,44,47,50,53,56,59,62,65,68,71-tetracosaoxatriheptacontane
M3050-100MG
M3050-25MG
TCIAM3050-100MGEA
8425
SEK
TCIAM3050-100MG
TCIAM3050-25MG
Methyl-PEG24-Azide ≥95.0%
Methyl-PEG24-Azide
Formel:
C₄₉H₉₉N₃O₂₄ MW: 1114,33 g/mol Storage Temperature: Frys |
CAS nummer:
2563873-82-3 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Elemental analysis (Nitrogen) | 3.50 to 4.00 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...