Methyl (Z)-3-(p-Toluenesulphonyloxy)but-2-enoate ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
Methyl (Z)-3-(Tosyloxy)but-2-enoate, (Z)-3-(p-Toluenesulphonyloxy)but-2-enoic acid methyl ester
M3043-1G
M3043-5G
TCIAM3043-1GEA
885
SEK
TCIAM3043-1G
TCIAM3043-5G
Methyl (Z)-3-(p-Toluenesulphonyloxy)but-2-enoate ≥98.0%
Methyl (Z)-3-(p-Toluenesulphonyloxy)but-2-enoate
Formel:
C₁₂H₁₄O₅S MW: 270,30 g/mol |
MDL Number:
MFCD30749294 CAS nummer: 1029612-18-7 |
Specifikation testresultat
Purity (GC) | min. 98.0 % |
Melting point | 63.0 to 67.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...