4-Methylumbelliferyl-2-acetamido-2-deoxy-β-D-glucopyranoside ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI

Synonyms: 4-Methylumbelliferyl 2-Acetamido-2-deoxy-beta-D-glucopyranoside, 4-Methylumbelliferyl N-Acetyl-beta-D-glucosaminide

M3030-250MG M3030-1G
TCIAM3030-250MGEA 855 SEK
TCIAM3030-250MG TCIAM3030-1G
4-Methylumbelliferyl-2-acetamido-2-deoxy-β-D-glucopyranoside ≥98.0% (by HPLC, total nitrogen)
4-Methylumbelliferyl-2-acetamido-2-deoxy-β-D-glucopyranoside
Formel: C₁₈H₂₁NO₈
MW: 379.365 g/mol
Storage Temperature: Frys
MDL Number: MFCD00065481
CAS nummer: 37067-30-4
EINECS: 253-333-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4fec1051e6d0b040d2ec79fe2d66baf937b4729d&vid=4fec1051e6d0b040d2ec79fe2d66baf937b4729d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D -29.0 to -33.0 deg (C=1, Pyridine)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR