Methyl-3-bromo-4-fluorobenzoate ≥98.0%
Leverantör: TCI
Varning
Synonyms: Methyl 3-bromo-4-fluorobenzoate
M3018-1G M3018-5G
TCIAM3018-1GEA 310 SEK
TCIAM3018-1G TCIAM3018-5G
Methyl-3-bromo-4-fluorobenzoate ≥98.0%
Methyl-3-bromo-4-fluorobenzoate
Formel: C₈H₆BrFO₂
MW: 233.04 g/mol
MDL Number: MFCD04972915
CAS nummer: 82702-31-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=118dbc846f45ddf87e7cd326f98903aa4ba945dd&vid=118dbc846f45ddf87e7cd326f98903aa4ba945dd&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %
Acid Value max. 1.0