6-Methoxyquinoline N-Oxide ≥98.0%
Leverantör: TCI
M2999-1G
M2999-5G
TCIAM2999-1GEA
1275
SEK
TCIAM2999-1G
TCIAM2999-5G
6-Methoxyquinoline N-Oxide ≥98.0%
6-Methoxyquinoline N-Oxide
Formel:
C₁₀H₉NO₂ |
MDL Number:
MFCD00009771 CAS nummer: 6563-13-9 |
|
Specifikation testresultat
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 108.0 to 112.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...