2-Methylbenzoic anhydride 98,0 GC, T

Leverantör: TCI
Varning

M2997-1G M2997-5G
TCIAM2997-1GEA 1275 SEK
TCIAM2997-1G TCIAM2997-5G
2-Methylbenzoic anhydride 98,0 GC, T
2-Methylbenzoic anhydride
Formel: C₁₆H₁₄O₃
MW: 254.29 g/mol
Kokpunkt: 221°C
MDL Number: MFCD00048077
CAS nummer: 607-86-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12d430326b081fc4130ea7f595992c0d35b48459&vid=12d430326b081fc4130ea7f595992c0d35b48459&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Morpholine Method) min. 98.0 %
Melting point 37.0 to 41.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR