Methyl benzenesulphinate ≥98.0%
Leverantör: TCI
M2971-5G
M2971-25G
M2971-1G
TCIAM2971-5GEA
1075
SEK
TCIAM2971-5G
TCIAM2971-25G
TCIAM2971-1G
Methyl benzenesulphinate ≥98.0%
Methyl benzenesulphinate
Formel:
C₇H₈O₂S Kokpunkt: 63°C Flampunkt: 92°C |
MDL Number:
MFCD00674051 CAS nummer: 670-98-4 |
|
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...