4-Methoxy-trans-stilbene ≥98.0%
Leverantör: TCI EUROPE
M2964-1G M2964-5G
TCIAM2964-1GEA 1025 SEK
TCIAM2964-1G TCIAM2964-5G
4-Methoxy-trans-stilbene ≥98.0%
4-Methoxy-trans-stilbene
Formel: C₁₅H₁₄O
MW: 210,28 g/mol
Kokpunkt: 126°C
MDL Number: MFCD00017178
CAS nummer: 1694-19-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f0a915a979a5e895b2742fe926e8ac562e20f8c&vid=5f0a915a979a5e895b2742fe926e8ac562e20f8c&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 134.0 to 138.0 deg-C