2-Methyl-5-nitrobenzaldehyde ≥98.0%

Leverantör: TCI
Varning

M2957-1G M2957-5G
TCIAM2957-1GEA 975 SEK
TCIAM2957-1G TCIAM2957-5G
2-Methyl-5-nitrobenzaldehyde ≥98.0%
2-Methyl-5-nitrobenzaldehyde
Formel: C₈H₇NO₃
MW: 165.1500 g/mol
Kokpunkt: 114°C
MDL Number: MFCD03840506
CAS nummer: 16634-91-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c612f5441088a0f0451a1f5629a20da13bece4cf&vid=c612f5441088a0f0451a1f5629a20da13bece4cf&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 53.0 to 57.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR