5-Methyl-2-(2H-1,2,3-triazol-2-yl)benzoic acid ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

M2935-1G M2935-5G
TCIAM2935-1GEA 510 SEK
TCIAM2935-1G TCIAM2935-5G
5-Methyl-2-(2H-1,2,3-triazol-2-yl)benzoic acid ≥98.0% (by HPLC, titration analysis)
5-Methyl-2-(2H-1,2,3-triazol-2-yl)benzoic acid
Formel: C₁₀H₉N₃O₂
MW: 203,197 g/mol
MDL Number: MFCD18382679
CAS nummer: 956317-36-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=048085d9f7cf7fbeab2ca2c81bd8f8ef2e405966&vid=048085d9f7cf7fbeab2ca2c81bd8f8ef2e405966&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 173.0 to 177.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR