1-Methoxy-4-methylnaphthalene ≥97.0%

Leverantör: TCI EUROPE

M2930-200MG M2930-1G
TCIAM2930-200MGEA 615 SEK
TCIAM2930-200MG TCIAM2930-1G
1-Methoxy-4-methylnaphthalene ≥97.0%
1-Methoxy-4-methylnaphthalene
Formel: C₁₂H₁₂O
MW: 172,23 g/mol
Kokpunkt: 155°C
MDL Number: MFCD00021472
CAS nummer: 24821-54-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ddf505f1151ed71057aaad1435b8c1759a6270c&vid=5ddf505f1151ed71057aaad1435b8c1759a6270c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR