Methyl-5-bromopyrimidine-2-carboxyate ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Methyl 5-Bromopyrimidine-2-carboxylate, 5-Bromopyrimidine-2-carboxylic acid methyll ester

M2910-1G M2910-200MG
TCIAM2910-1GEA 1100 SEK
TCIAM2910-1G TCIAM2910-200MG
Methyl-5-bromopyrimidine-2-carboxyate ≥98.0%
Methyl-5-bromopyrimidine-2-carboxyate
Formel: C₆H₅BrN₂O₂
MW: 217,02 g/mol
Smältpunkt: 149°C
MDL Number: MFCD11111606
CAS nummer: 89581-38-4

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR