2-Morpholinoethyl methacrylate ≥95.0% stabilised

Leverantör: TCI EUROPE
Varning

Synonyms: Methacrylic acid 2-Morpholinoethyl ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

M2901-1G M2901-5G
TCIAM2901-1GEA 430 SEK
TCIAM2901-1G TCIAM2901-5G
2-Morpholinoethyl methacrylate ≥95.0% stabilised
2-Morpholinoethyl methacrylate
Formel: C₁₀H₁₇NO₃
MW: 199,25 g/mol
Kokpunkt: 81°C
MDL Number: MFCD00080577
CAS nummer: 2997-88-8
EINECS: 221-069-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69bf3271fa183b3d69792889b8ac7f4c005b15f6&vid=69bf3271fa183b3d69792889b8ac7f4c005b15f6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light red clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR