(7-Methoxy-1-naphthyl)acetic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 7-Methyl-1-naphthyl acetic acid, 7-Methoxy-1-naphthaleneacetic acid, 2-(7-Methoxynaphthalen-1-yl)acetic acid, (7-Methoxy-1-naphthyl)acetic acid

M2888-1G M2888-5G
TCIAM2888-1GEA 655 SEK
TCIAM2888-1G TCIAM2888-5G
(7-Methoxy-1-naphthyl)acetic acid ≥98.0% (by GC, titration analysis)
(7-Methoxy-1-naphthyl)acetic acid
Formel: C₁₃H₁₂O₃
MW: 216,24 g/mol
MDL Number: MFCD12756806
CAS nummer: 6836-22-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27be9e59116927cc6d8c161834e1ea96f3671346&vid=27be9e59116927cc6d8c161834e1ea96f3671346&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 155.0 to 159.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR