4-Methoxy-6-methyl-2H-pyran-2-one ≥98.0%
Leverantör: TCI EUROPE
M2881-1G
M2881-200MG
TCIAM2881-1GEA
1175
SEK
TCIAM2881-1G
TCIAM2881-200MG
4-Methoxy-6-methyl-2H-pyran-2-one ≥98.0%
4-Methoxy-6-methyl-2H-pyran-2-one
Formel:
C₇H₈O₃ MW: 140,138 g/mol |
MDL Number:
MFCD00006640 CAS nummer: 672-89-9 EINECS: 211-600-9 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 86.0 to 90.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...