2'-Methoxy-1'-acetonaphthone ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1-Acetyl-2-methoxynaphthalene, 1-(2-Methoxynaphthalen-1-yl)ethanone
M2875-1G
M2875-5G
TCIAM2875-1GEA
1025
SEK
TCIAM2875-1G
TCIAM2875-5G
2'-Methoxy-1'-acetonaphthone ≥98.0%
2'-Methoxy-1'-acetonaphthone
Formel:
C₁₃H₁₂O₂ MW: 200,24 g/mol Kokpunkt: 158°C |
MDL Number:
MFCD11553477 CAS nummer: 5672-94-6 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 56.0 to 60.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...